Invitrogen™ RPS6KA1 (RSK1) Recombinant Human Protein DFS Item
RPS6KA1 (RSK1) is a growth-factor regulated serine/threonine kinase that is involved in the MAPK cascade.
Manufacturer:  Invitrogen™ LSPV4049
 View more versions of this product
Catalog No. PV4049
Description
Description
RPS6KA1 (RSK1) is thought to control cellular proliferation and differentiation through the phosphorylation of transcription factors.
Specifications
Specifications
NP_002944
Frozen
88.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
RSK (RPS6K) Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
RPS6KA1 (RSK1)
Baculovirus
Human
His Tag
Documents