Please login to your online account to display your discounted pricing

Roxindole hydrochloride, Tocris

Dopamine D2 autoreceptor agonist; Also has affinity for D3, D4, 5-HT1A receptors and the 5-HT transporter

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Roxindole hydrochloride
Assay Percent Range >98%
Molecular Formula C23H26N2O·HCl
Formula Weight 382.93
Synonym 3-[4-(3, 6-Dihydro-4-phenyl-1(2H)-pyridinyl)butyl]-1H-indol-5-ol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

155950

 
r & d systems
RDS155950
50mg Each for $844.73

155910

 
r & d systems
RDS155910
10mg Each for $201.36
Description & Specifications

Specifications

Formula Weight 382.93
Quantity 50mg
CAS 108050-82-4
Chemical Name or Material Roxindole hydrochloride
Assay Percent Range >98%
Molecular Formula C23H26N2O·HCl
Synonym 3-[4-(3, 6-Dihydro-4-phenyl-1(2H)-pyridinyl)butyl]-1H-indol-5-ol hydrochloride
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C