missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Roxatidine Acetate Hydrochloride 98.0+%, TCI America™

Manufacturer:  Tci America R01781G

 View more versions of this product

Catalog No. R01781G



Specifications

Specifications

Roxatidine Acetate Hydrochloride
148°C
93793-83-0
MFCD00941429
FEWCTJHCXOHWNL-UHFFFAOYSA-N
[2-oxo-2-[3-[3-(piperidin-1-ylmethyl)phenoxy]propylamino]ethyl] acetate;hydrochloride
56704
Crystal-Powder at 20°C
White
1g
C19H29ClN2O4
2-Acetoxy-N-[3-[3-(1-piperidinylmethyl)phenoxy]propyl]acetamide Hydrochloride
CC(=O)OCC(=O)NCCCOC1=CC=CC(=C1)CN2CCCCC2.Cl
384.901
384.90
≥98.0% (HPLC,T)

Chemical Identifiers

93793-83-0
384.901
FEWCTJHCXOHWNL-UHFFFAOYSA-N
56704
CC(=O)OCC(=O)NCCCOC1=CC=CC(=C1)CN2CCCCC2.Cl
C19H29ClN2O4
MFCD00941429
2-Acetoxy-N-[3-[3-(1-piperidinylmethyl)phenoxy]propyl]acetamide Hydrochloride
[2-oxo-2-[3-[3-(piperidin-1-ylmethyl)phenoxy]propylamino]ethyl] acetate;hydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : AC3855000

TSCA : No

Documents