Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Coated Clear Straight Sided Round Bottles with White Polypropylene Pulp/Vinyl Cap
Reduce breakage and lab mishaps. Fisherbrand™ Safety Coated Clear Straight Sided Round Bottles with White Polypropylene Pulp/Vinyl Cap are ideal for soil sampling and environmental applications.
$86.50 - $347.33
Specifications
Closure Color White
Closure Material Polypropylene
Liner Material Pulp/Vinyl
Color Clear
Material Safety Coated Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity Neck Finish Price Quantity    
02-911-824
fisherbrand™
264029
16 oz., 473mL 89-400 Case of 12 Each for $347.33
02-911-825
fisherbrand™
264047
8 oz., 250mL 70-400 Case of 12 Each for $86.50
Description & Specifications
Specifications
Closure Color White
Closure Material Polypropylene
Liner Material Pulp/Vinyl
Color Clear
Material Safety Coated Glass
Mouth Wide
Shape Straight Sided
Closure Included Yes
  • Plastisol coating contains substance in bottle long enough to allow for proper disposal if the glass breaks
  • Straight sided walls allow for the complete removal of contents
  • Features polypropylene polypropylene pulp/vinyl cap