Please login to your online account to display your discounted pricing
Fisherbrand™ Clear Straight Sided Round Bottles with White Polypropylene PE Foam/PTFE Cap
Multipurpose jars for maximum content visibility. Fisherbrand™ Clear Straight Sided Round Bottles with White Polypropylene PE Foam/PTFE Cap are designed without shoulders for maximum storage.
$163.17 - $1,066.07
Specifications
Closure Color White
Closure Material Polypropylene
Closure Liner PTFE
Color Clear
Material Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Neck Finish Price Quantity    
02-911-791
fisherbrand™
264033
60mL 53-400 Case of 144 Each for $513.83
02-911-912
fisherbrand™
264031
30mL 43-400 Case of 48 Each for $163.17
02-912-306
fisherbrand™
264030
30mL 43-400 Case of 384 Each for $1,066.07
Description & Specifications
Specifications
Closure Color White
Closure Material Polypropylene
Closure Liner PTFE
Color Clear
Material Glass
Mouth Wide
Closure Included Yes
Available certified cleaned per EPA guidelines for semi-volatiles, pesticides, PBCs and metals. A certificate of analysis is attached to each lot. Safety coated bottles are ideal for field sampling applications. The shatter resistant coating reduces the chance of breakage in the event the container is dropped. If the bottle does break, the coating will safely contain the contents long enough for disposal. The non-slip coating also allows greater ease of stability in handling containers, whether wet or dry.
  • Straight sided walls allow for the complete removal of contents
  • Not autoclavable
  • Bottles are packed with caps attached Ideal for soil sampling and environmental applications.