Please login to your online account to display your discounted pricing
Fisherbrand™ Round Safety Coated Graduated Clear Soda Lime Glass Wide Mouth Packer Bottles
$183.65 - $183.65
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    
02-911-457
fisherbrand™
W014154
500mL Case of 24 Each for $183.65
Description & Specifications
Containers and caps which are 'custom cleaned' are cleaned of visible particulates using deionised water. This is non-certified - no documentation is attached for thisContainers are which are 'certified clean' are cleaned as per EPA guidelines for semi-volatiles, pesticides, PCBs and metals. A certificate of analysis is attached to each lot. Safety coated bottles are ideal for field sampling applications. The shatter resistant coating reduces the chance of breakage in the event the container is dropped. If the bottle does break, the coating will safely contain the contents long enough for disposal. The non-slip bottles are packed with caps attached.
  • Wide mouth bottles allow for easy accessibility for removal of contents
  • Available in clear and amber Ideal for storage of dry powders.