Please login to your online account to display your discounted pricing
Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles without Cap
General use bottles ideal when UV protection is important. Fisherbrand™ Safety Coated Amber Wide Mouth Packer Bottles without Cap absorbs all wavelengths in the ultraviolet spectrum.
$181.73 - $199.53
Specifications
Closure Screw
Color Amber
Mouth Style Wide
Closure Included No
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Neck Finish Price Quantity    
02-911-907
fisherbrand™
264024
1L Case of 12 Each for $181.73
02-912-317
fisherbrand™
264007
8.5oz., 250mL 45-400 Case of 24 Each for $199.53
Description & Specifications
Specifications
Closure Screw
Color Amber
Mouth Style Wide
Closure Included No
  • Features narrow screw neck opening, curved shoulders, and a round body
  • Offers minimal chemical interference with contents
  • Ensure integrity for long term storage and protection for UV-sensitive contents