Please login to your online account to display your discounted pricing

Corning™ Round Ice Bucket with Lid, 4L


Multi-purpose EVA buckets that provide a non-toxic alternative to conventional ice buckets

Manufacturer: corning™  C432125

 View more versions of this product

Catalog No. 07-210-125

  • / Case
 

Description & Specifications

Specifications

No. per Case 1
Capacity 4L
Color Orange
Dimensions (L x W x H) 18.4 x 26cm