Please login to your online account to display your discounted pricing

Corning™ Round Ice Bucket with Lid, 2.5L


Multi-purpose EVA buckets that provide a non-toxic alternative to conventional ice buckets

Manufacturer: corning™  C432134

 View more versions of this product

Catalog No. 07-210-134

  • / Case
 

Description & Specifications

Specifications

Capacity 2.5L
Color Pink
Dimensions (L x W x H) 12.1 x 24.1cm