Please login to your online account to display your discounted pricing
Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene PE Foam/PTFE Cap
Ideal general purpose bottle. Fisherbrand™ Clear Standard Wide Mouth Bottles with White Polypropylene PE Foam/PTFE Cap used for pharmaceutical, chemical, environmental, and food packaging and storing applications.
$53.00 - $120.87
Specifications
Closure Color White
Color Clear
Material Glass
Liner Material PE Foam/PTFE
Mouth Wide
View More Specs
Products
Catalog Number Mfr. No. Quantity Capacity (Metric) Neck Finish Price Quantity    
02-911-767
fisherbrand™
262936
24/Cs. 480mL 63-400 Case of 24 Each for $120.87
02-911-770
fisherbrand™
262931
24/Cs. 240mL 58-400 Case of 24 Each for $89.30
02-911-768
fisherbrand™
262930
24/Cs. 120mL 48-400 Case of 24 Each for $53.00
02-911-769
fisherbrand™
262939
6/Cs. 1920mL 83-400 Case of 6 Each for $61.70
02-911-913
fisherbrand™
262938
12/Cs. 960mL 70-400 Case of 12 Each for $82.27
02-911-766
fisherbrand™
262941
4/Cs. 3785mL 89-400 Case of 4 Each for $56.03
Description & Specifications
Specifications
Closure Color White
Color Clear
Material Glass
Liner Material PE Foam/PTFE
Mouth Wide
  • Features wide mouth for easy transfering of contents
  • Available in capacities ranging from 120 to 3840mL