Please login to your online account to display your discounted pricing
Fisherbrand™ Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap
Ideal general purpose bottle. Fisherbrand™ Clear Boston Round Bottles with White Polypropylene Pulp/Vinyl Cap ensure integrity for long term storage.
$221.53 - $334.70
Specifications
Closure Color White
Closure Material Polypropylene
Closure Liner Poly/Vinyl
Color Clear
Mouth Narrow
View More Specs
Products
Catalog Number Mfr. No. Neck Finish Capacity (Metric) Price Quantity    
02-911-730
fisherbrand™
263924
22-400 120mL Case of 160 Each for $239.67
02-911-732
fisherbrand™
263937
24-400 240mL Case of 108 Each for $221.53
02-911-790
fisherbrand™
263898
20-400 60mL Case of 288 Each for $334.70
Description & Specifications
Specifications
Closure Color White
Closure Material Polypropylene
Closure Liner Poly/Vinyl
Color Clear
Mouth Narrow
Shape Boston Round
  • Features narrow screw neck opening, curved shoulders and round body