Rotronic™ Humidity standards
For on-site calibration and adjustment of ROTRONIC probes with handheld instrument
Manufacturer:  Rotronic™ 5010EA05SCS1
 View more versions of this product
Catalog No. 11-650-3
Includes: SCS traceable calibration certificate, textile pads, Calibration instructions
Description
Description
With the humidity standards, a calibration device and HW4 software, running on a PC this is easily achieved.
  • Ampoules contain unsaturated salt solutions
  • Simple and safe on-site calibration method
  • Unlimited storage life
Specifications
Specifications
Rotronic instrument
SCS traceable calibration certificate, textile pad, calibration instruction
Humidity Standard
5%
On site calibration and adjustment of rotronic probe