Thermo Scientific™ Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Expand individual bucket capacity or maximize total blood bags per spin with choice of buckets for Thermo Scientific™ HAEMAFlex™ blood banking rotors.
$2,227.48 - $5,976.17
Catalog Number | Mfr. No. | Description | For Use With (Equipment) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | For Use With (Equipment) | Price | Quantity | ||||
75-003-835
|
Thermo Scientific™
75003835 |
Oval, Single Blood Bag Bucket, with Dura-Coat (set of 2) | HAEMAFlex 6/8 swinging bucket rotors |
Pack of 2 Each for $3,525.75
|
|
75-003-834
|
Thermo Scientific™
75003834 |
Oval, Single, Blood Bag, Bucket, 6/8 Swinging Bucket Rotor | HAEMAFlex 6/8 swinging bucket rotors |
Pack of 2 Each for $2,227.48
|
|
75-003-836
|
Thermo Scientific™
75003836 |
Oval, Single Blood Bag Bucket, with Dura-Coat (set of 2) | HAEMAFlex 6/8 swinging bucket rotors |
Pack of 2 Each for $2,328.65
|
|
75-003-846
|
Thermo Scientific™
75003846 |
12/16 Swinging Bucket Rotors | HAEMAFlex 12/16 swinging bucket rotors |
Pack of 2 Each for $5,567.77
|
|
75-003-865
|
Thermo Scientific™
75003865 |
Higher capacity, 12/16 Swing Bucket Rotors | HAEMAFlex 12/16 swinging bucket rotors |
Pack of 2 Each for $5,976.17
|
|
75-003-864
|
Thermo Scientific™
75003864 |
Oval, Single Blood Bag Bucket, with Dura-Coat (set of 2) | HAEMAFlex 6/8 swinging bucket rotors |
Pack of 2 Each for $2,497.25
|
|
Description
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.For high-throughput blood processing needs, the HAEMAFlex 12 swinging bucket rotor provides 12 x 500mL capacity or upgrade your centrifuge capacity to 16 x 500mL blood bags with the HAEMAFlex 16 rotor and the same bucket selections.
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.