missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rotenone, ≥97%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0215015401
Specifications
Rotenone | |
1g | |
≥97% | |
--cis-rotenone, --rotenone, barbasco, cube root, dactinol, derrin, derris, paraderil, rotenone, tubatoxin | |
CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC | |
6758 | |
394.4 |
Beige to White | |
83-79-4 | |
C23H22O6 | |
JUVIOZPCNVVQFO-HBGVWJBISA-N | |
394.423 | |
CHEBI:28201 | |
Powder |
Chemical Identifiers
83-79-4 | |
394.423 | |
--cis-rotenone, --rotenone, barbasco, cube root, dactinol, derrin, derris, paraderil, rotenone, tubatoxin | |
CHEBI:28201 |
C23H22O6 | |
JUVIOZPCNVVQFO-HBGVWJBISA-N | |
6758 | |
CC(=C)C1CC2=C(O1)C=CC3=C2OC4COC5=CC(=C(C=C5C4C3=O)OC)OC |
Promotions
-
Spend $500, Get $250 on MP Biomedicals Products
Buy $500 or more in MP Biomedicals products and get $250 more to spend on products at no additional cost.: 06/30/21