missing translation for 'onlineSavingsMsg'
Learn More

Rosuvastatin, calcium salt, 98%

CAS: 147098-20-2 | C22H28FN3O61/2Ca | 500.58 g/mol

$273.95 - $273.95

Chemical Identifiers

CAS 147098-20-2
Molecular Formula C22H28FN3O61/2Ca
Molecular Weight (g/mol) 500.58
InChI Key LQFSLFAVBQEFBH-LSMDDVBQSA-N
Synonym 3s,5r-rosuvastatin calcium salt, ent-rosuvastatin
PubChem CID 131673834
IUPAC Name calcium;(E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid;molecular hydrogen
SMILES [HH].[HH].CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.[Ca]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC456232500
SDS View Documents
Thermo Scientific Chemicals
456232500
250 mg
Each for $273.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

147098-20-2
500.58
3s,5r-rosuvastatin calcium salt, ent-rosuvastatin
calcium;(E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid;molecular hydrogen
C22H28FN3O61/2Ca
LQFSLFAVBQEFBH-LSMDDVBQSA-N
131673834
[HH].[HH].CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.[Ca]

Specifications

147098-20-2
97.5% min. (HPLC)
250 mg
LQFSLFAVBQEFBH-LSMDDVBQSA-N
calcium;(E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid;molecular hydrogen
131673834
98%
Authentic
C22H28FN3O61/2Ca
3s,5r-rosuvastatin calcium salt, ent-rosuvastatin
[HH].[HH].CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.[Ca]
500.58
500.58
Rosuvastatin, calcium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of damaging the unborn child.
May cause harm to breast-fed children.

GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid contact during

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only