missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rosiglitazone 98.0+%, TCI America™

Manufacturer:  Tci America R01061G

 View more versions of this product

Catalog No. R01061G



Specifications

Specifications

Rosiglitazone
C18H19N3O3S
YASAKCUCGLMORW-UHFFFAOYSA-N
5-[[4-[2-[methyl(pyridin-2-yl)amino]ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
77999
357.43
≥98.0% (HPLC,T)
White-Yellow
122320-73-4
MFCD00871760
CN(CCOC1=CC=C(C=C1)CC2C(=O)NC(=O)S2)C3=CC=CC=N3
357.428
CHEBI:50122
Crystal-Powder at 20°C
156°C
1g

Chemical Identifiers

122320-73-4
357.428
YASAKCUCGLMORW-UHFFFAOYSA-N
CHEBI:50122
CN(CCOC1=CC=C(C=C1)CC2C(=O)NC(=O)S2)C3=CC=CC=N3
C18H19N3O3S
MFCD00871760
77999
5-[[4-[2-[methyl(pyridin-2-yl)amino]ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
Safety and Handling

Safety and Handling

RTECSNumber : XJ5813850

TSCA : No

Recommended Storage : Refrigerator

Documents