missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rosiglitazone 98.0+%, TCI America™

$67.78 - $239.50

Chemical Identifiers

CAS 122320-73-4
Molecular Formula C18H19N3O3S
Molecular Weight (g/mol) 357.428
MDL Number MFCD00871760
InChI Key YASAKCUCGLMORW-UHFFFAOYSA-N
PubChem CID 77999
ChEBI CHEBI:50122
IUPAC Name 5-[[4-[2-[methyl(pyridin-2-yl)amino]ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
SMILES CN(CCOC1=CC=C(C=C1)CC2C(=O)NC(=O)S2)C3=CC=CC=N3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
R0106200MG View Documents Tci America
R0106200MG
200mg Each for $67.78
 
R01061G View Documents Tci America
R01061G
1g Each for $239.50
 
Specifications

Chemical Identifiers

122320-73-4
357.428
YASAKCUCGLMORW-UHFFFAOYSA-N
CHEBI:50122
CN(CCOC1=CC=C(C=C1)CC2C(=O)NC(=O)S2)C3=CC=CC=N3
C18H19N3O3S
MFCD00871760
77999
5-[[4-[2-[methyl(pyridin-2-yl)amino]ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione

Specifications

122320-73-4
MFCD00871760
CN(CCOC1=CC=C(C=C1)CC2C(=O)NC(=O)S2)C3=CC=CC=N3
357.428
CHEBI:50122
Crystal-Powder at 20°C
156°C
200mg
C18H19N3O3S
YASAKCUCGLMORW-UHFFFAOYSA-N
5-[[4-[2-[methyl(pyridin-2-yl)amino]ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
77999
357.43
≥98.0% (HPLC,T)
White-Yellow
Rosiglitazone
Safety and Handling

Safety and Handling

RTECSNumber : XJ5813850

TSCA : No

Recommended Storage : Refrigerator