ROS1 Recombinant Human Protein DFS Item
ROS1 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3814
Catalog No. PV3814
Description
Description
ROS1 is a tyrosine kinase that functions as a growth or differentiation factor receptor.
Specifications
Specifications
Kinase assay
Baculovirus
Receptor tyrosine kinases
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
79.6kDa
80% by SDS-PAGE
–80°C
1 Each
ROS1 Recombinant Human Protein
SEV
ROS1 Recombinant Human Protein
Human
NP_002935
Kinase and inhibitors
0.36mg/mL
6098
ROS1 Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
ROS1, ROS, MCF4
ROS1 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 670μg/mL Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C
Amino acids 1883-2347
4806nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.104μg/mL
GST Tag
SDS
Documents
For Research Use Only. Not for use in diagnostic procedures.