Invitrogen™ ROS1 Recombinant Human Protein DFS Item
ROS1 is a tyrosine protein kinase.
Manufacturer:  Invitrogen™ LSPV4120
 View more versions of this product
Catalog No. PV4120
Description
Description
ROS1 functions as a growth or differentiation factor receptor.
Specifications
Specifications
NP_002935
Frozen
79.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
SEV Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
ROS1, ROS, MCF4
Baculovirus
Human
GST Tag
Documents