Please login to your online account to display your discounted pricing

ROS 234 dioxalate, Tocris

Potent H3 antagonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material ROS 234 dioxalate
Assay Percent Range >99%
Molecular Formula C13H15N5·2C2H2O4
Formula Weight 421.37
Synonym N-[3-(1H-Imidazol-4-yl)propyl]-1H-benzimidazol-2-amine dioxalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

203410

 
r & d systems
RDS203410
10mg Each for $181.19

203450

 
r & d systems
RDS203450
50mg Each for $762.58
Description & Specifications

Specifications

Formula Weight 421.37
Quantity 10mg
CAS 184576-87-2
Chemical Name or Material ROS 234 dioxalate
Assay Percent Range >99%
Molecular Formula C13H15N5·2C2H2O4
Synonym N-[3-(1H-Imidazol-4-yl)propyl]-1H-benzimidazol-2-amine dioxalate
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature