Invitrogen™ ROR2 Recombinant Human Protein DFS Item
ROR2 (receptor tyrosine kinase-like orphan receptor 2) is a type I transmembrane protein that belongs to the ROR subfamily of cell surface receptors.
Manufacturer:  Invitrogen™ LSPV4179
 View more versions of this product
Catalog No. PV4179
Description
Description
ROR2 is thought to be involved in the early formation of the chondrocytes and cartilage and growth plate development.
Specifications
Specifications
NP_004551
Frozen
85.2kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
ROR Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
ROR2
Baculovirus
Human
GST Tag
Documents