missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ROR gamma (t) Rat anti-Mouse, PE, Clone: B2D, eBioscience™

Rat Monoclonal Antibody

$209.88 - $550.44

Specifications

Antigen ROR gamma (t)
Clone B2D
Host Species Rat
Gene Alias Retinoid-Related Orphan Receptor gamma
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245520 Life Technologies
LS12698182
100μg Each for $550.44
501129397 Life Technologies
LS12698180
25μg Each for $209.88
Specifications

Description

The B2D monoclonal antibody reacts with the mouse ROR gamma (t) protein. ROR gamma is a member of the retinoic acid-related orphan receptor (ROR) family, which also includes ROR alpha and ROR beta. ROR family proteins are ligand-dependent transcription factors that play roles in multiple physiological processes. ROR gamma is expressed in several tissues including liver, lung, muscle, heart and kidney. Furthermore, it was discovered that alternative transcription results in the expression of an isoform, ROR gamma t, which is expressed exclusively in cells of the lymphoid compartment, namely CD4+CD8+ “double-positive” thymoyctes, Th17 cells of the periphery and lymphoid tissue inducer (Lti) cells of lymphoid organs.

The ROR gamma t isoform differs from ROR gamma by three unique amino acids at the amino terminus. Therefore, the B2D antibody will react with both the ROR gamma and ROR gamma t isoforms.
Description & Specifications

Specifications

ROR gamma (t)
Rat
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
B2D
Retinoid-Related Orphan Receptor gamma
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer