Roots and Stems Prepared Slides
Slides give magnified view of Gymnosperms and Angiosperms for detailed study.
Supplier: Customer Channels Group (Ccg) 142FL
Description
- Pine cross section provides excellent view of secondary xylem for discussing formation of annual rings
- Determine the difference between monocot and dicot roots, stems, and leaves
- Flower reproductive parts such as pollen and pollen tubes are clearly visible
Specifications
Allium, Root Tip L.s. | |
Roots and Stems | |
Prepared Slide |