Please login to your online account to display your discounted pricing

Rolipram, Tocris


PDE4 inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS090510

 View more versions of this product

Catalog No. 090510

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 275.35
Chemical Name or Material Rolipram
Assay Percent Range >98%
Molecular Formula C16H21NO3
CAS 61413-54-5
Synonym 4-(3-(Cyclopentyloxy)-4-methoxyphenyl)pyrrolidin-2-one
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Rolipram
Assay Percent Range >98%
Molecular Formula C16H21NO3
Formula Weight 275.35
Synonym 4-(3-(Cyclopentyloxy)-4-methoxyphenyl)pyrrolidin-2-one
Accessories

Store at Room Temperature