Please login to your online account to display your discounted pricing

Rolipram, Tocris

PDE4 inhibitor

$142.08 - $594.43

Chemical Identifiers

Chemical Name or Material Rolipram
Assay Percent Range >98%
Molecular Formula C16H21NO3
Formula Weight 275.35
Synonym 4-(3-(Cyclopentyloxy)-4-methoxyphenyl)pyrrolidin-2-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

090550

 
r & d systems
RDS090550
50mg Each for $594.43

090510

 
r & d systems
RDS090510
10mg Each for $142.08
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 275.35
Chemical Name or Material Rolipram
Assay Percent Range >98%
Molecular Formula C16H21NO3
CAS 61413-54-5
Synonym 4-(3-(Cyclopentyloxy)-4-methoxyphenyl)pyrrolidin-2-one
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature