missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rolipram, 98%

$60.14 - $60.14

Specifications

Description White to off-white
Format Solid
Assay 98%
CAS 61413-54-5
Molecular Formula C16H21NO3
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67177LB0 Alfa Aesar™
J67177LB0
5mg Each for $60.14
AAJ67177MA Alfa Aesar™
J67177MA
10mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67177MB Alfa Aesar™
J67177MB
25mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Specifications

White to off-white
98%
C16H21NO3
4-(3-Cyclopentyloxy-4-methoxyphenyl)-2-pyrrolidinone; ZK 62711
Solid
61413-54-5
275.35g/mol