missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

American Educational Products Rocks and Minerals of the U.S. Collections

Choose from three sets: Introductory, Basic, and the United States collection.

$111.37 - $111.37

Specifications

Specimen Size 0.75in.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S45047
View Documents
American Educational Products
S45047
100 Specimens
Each for $111.37
 
Description

Description

Introductory Collection of Rocks and Minerals.

  • 15 common and colorful rocks and minerals
  • Size (approx.): 0.75 in.
Basic Rocks and Minerals Collection.
  • 35 rocks and rock-forming minerals
  • Displays igneous, metamorphic, and sedimentary rocks, plus the minerals that make up those rocks
  • Size (approx.): 0.75 in.
Rocks, Minerals and Gemstones of the United States Collection.
  • 64 mineral specimens following Dana's System of Mineralogy
  • 36 igneous, sedimentary, and metamorphic rocks
  • Includes study guide
  • Size (approx.): 0.75 in.

Specifications

Specifications

0.75in.
SDS
Product Certifications