Invitrogen™ ROCK2 Recombinant Human Protein DFS Item
ROCK2 is the predominant ROCK isoform in skeletal muscle.
Manufacturer:  Invitrogen™ LSPV4048
 View more versions of this product
Catalog No. PV4048
Description
Description
ROCK2 is an important regulator downstream of Rho GTPases.
Specifications
Specifications
NP_004841
Frozen
90.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
DMPK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
ROCK2
Baculovirus
Human
GST Tag
Documents