Please login to your online account to display your discounted pricing

Rock and Mineral Test Kit


Test to distinguish between rocks and minerals.

Manufacturer: fisher science education  AZ-20-3933

 View more versions of this product

Catalog No. S90621

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Packaging Quantity Set of 5
For Use With (Application) Used to conduct a series of tests to distinguish between rocks and minerals
Product Type Rock and mineral test kit
Sufficient For 5
Includes Six specimens, streak plate, plastic dropper bottle, magnet and instructions