Please login to your online account to display your discounted pricing

Rock and Mineral Test Kit


Test to distinguish between rocks and minerals.

Manufacturer: fisher science education  AZ-20-3931

 View more versions of this product

Catalog No. S90620

  • / Each
 

Description & Specifications

Specifications

Packaging Quantity 1
Sufficient For 1
Product Type Rock and mineral test kit
Includes Six specimens, streak plate, plastic dropper bottle, magnet and instructions
For Use With (Application) Used to conduct a series of tests to distinguish between rocks and minerals