Please login to your online account to display your discounted pricing

American Educational Products Rock Cycle Classroom Project


Millions of years of geological processes captured in one chart.

Manufacturer: american educational products  2720EMD

 View more versions of this product

Catalog No. S45054

  • / Each
 

Description & Specifications

Specifications

Grade 6 to 12
For Use With (Application) Enable students to examine and identify individual rock and mineral specimens
Product Type Rock cycle classroom project
Includes Wall chart, 30 students charts, 30 specimens of each rock on the chart, test kit and a study guide