Please login to your online account to display your discounted pricing

Ro 90-7501, Tocris

Inhibitor of Aβ42 fibril formation

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material Ro 90-7501
Assay Percent Range >99%
Molecular Formula C20H16N6
Formula Weight 340.39
Synonym 2'-(4-Aminophenyl)-[2, 5'-bi-1H-benzimidazol]-5-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

240810

 
r & d systems
240810
10mg Each for $205.70

240850

 
r & d systems
240850
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 340.39
Quantity 10mg
Chemical Name or Material Ro 90-7501
Assay Percent Range >99%
Molecular Formula C20H16N6
CAS 293762-45-5
Synonym 2'-(4-Aminophenyl)-[2, 5'-bi-1H-benzimidazol]-5-amine
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C