Please login to your online account to display your discounted pricing

Ro 8-4304 hydrochloride, Tocris

Selective NR2B antagonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material Ro 8-4304 hydrochloride
Assay Percent Range >99%
Molecular Formula C21H23FN2O3·HCl
Formula Weight 406.88
Synonym 4-[3-[4-(4-Fluorophenyl)-1,2,3,6-tetrahydro-1(2H)-pyridinyl]-2-hydroxypropoxy]benzamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

334310

 
r & d systems
RDS334310
10mg Each for $205.70

334350

 
r & d systems
RDS334350
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 406.88
Quantity 10mg
Assay Percent Range >99%
Molecular Formula C21H23FN2O3·HCl
CAS 1312991-77-7
Synonym 4-[3-[4-(4-Fluorophenyl)-1,2,3,6-tetrahydro-1(2H)-pyridinyl]-2-hydroxypropoxy]benzamide hydrochloride
Chemical Name or Material Ro 8-4304 hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature