Please login to your online account to display your discounted pricing

Ro 67-7476, Tocris™

Positive allosteric modulator of mGlu1

$233.76 - $963.39

Chemical Identifiers

Chemical Name or Material Ro 67-7476
Assay Percent Range >99%
Molecular Formula C17H18FNO2S
Formula Weight 319.39
Synonym (2S)-2-(4-Fluorophenyl)-1-[(4-methylphenyl)sulfonyl]-pyrrolidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

434610

 
r & d systems
RDS434610
10mg Each for $233.76

434650

 
r & d systems
RDS434650
50mg Each for $963.39
Description & Specifications

Specifications

Formula Weight 319.39
Quantity 10mg
Chemical Name or Material Ro 67-7476
Assay Percent Range >99%
Molecular Formula C17H18FNO2S
CAS 298690-60-5
Synonym (2S)-2-(4-Fluorophenyl)-1-[(4-methylphenyl)sulfonyl]-pyrrolidine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature