Please login to your online account to display your discounted pricing

Ro 64-5229, Tocris

Selective, non-competitive mGlu2 antagonist

$246.52 - $1,023.37

Chemical Identifiers

Chemical Name or Material Ro 64-5229
Assay Percent Range >99%
Molecular Formula C17H19Cl2N3O
Formula Weight 352.26
Synonym (Z)-1-[2-Cycloheptyloxy-2-(2,6-dichlorophenyl)ethenyl]-1H-1,2,4-triazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

291310

 
r & d systems
291310
10mg Each for $246.52

291350

 
r & d systems
291350
50mg Each for $1,023.37
Description & Specifications

Specifications

Formula Weight 352.26
Quantity 10mg
Chemical Name or Material Ro 64-5229
Assay Percent Range >99%
Molecular Formula C17H19Cl2N3O
CAS 246852-46-0
Synonym (Z)-1-[2-Cycloheptyloxy-2-(2,6-dichlorophenyl)ethenyl]-1H-1,2,4-triazole
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C