Please login to your online account to display your discounted pricing

Ro 51, Tocris™

Potent P2X3, P2X2/3 antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material Ro 51
Assay Percent Range >98%
Molecular Formula C17H23IN4O4
Formula Weight 474.29
Synonym 2-[[4-Amino-5-[5-iodo-4-methoxy-2-(1-methylethyl)phenoxy]-2, 4-pyrimidinyl]amino]-1, 3-propanediol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

439150

 
r & d systems
439150
50mg Each for $1,050.42

439110

 
r & d systems
439110
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 474.29
Quantity 50mg
Chemical Name or Material Ro 51
Synonym 2-[[4-Amino-5-[5-iodo-4-methoxy-2-(1-methylethyl)phenoxy]-2, 4-pyrimidinyl]amino]-1, 3-propanediol
Assay Percent Range >98%
CAS 1050670-85-3
Molecular Formula C17H23IN4O4
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C