missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RO-3306 95.0+%, TCI America™

Manufacturer:  TCI America R020110MG

 View more versions of this product

Catalog No. R020110MG



Specifications

Specifications

RO-3306
10mg
C18H13N3OS2
cdk1 inhibitor iv, ro-3306, 5-6-quinolinylmethylene-2-2-thienylmethyl amino-4 5h-thiazolone, 5z-5-quinolin-6-ylmethylidene-2-thiophen-2-ylmethyl amino-1,3-thiazol-4-one, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethylamino thiazol-4 5h-one, ro hplc, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethyl amino thiazol-4 5h-one, 2-thiophen-2-ylmethyl-amino-5-1-quinolin-6-yl-meth-z-ylidene-thiazol-4-one, 5z-5-quinolin-6-yl methylidene-2-thiophen-2-yl methyl amino-4,5-dihydro-1,3-thiazol-4-one
C1=CC2=C(C=CC(=C2)C=C3C(=O)N=C(S3)NCC4=CC=CS4)N=C1
351.442
351.44
≥95.0% (HPLC)
Undesignated
872573-93-8
MFCD17392573
XOLMRFUGOINFDQ-YBEGLDIGSA-N
(5Z)-5-(quinolin-6-ylmethylidene)-2-(thiophen-2-ylmethylamino)-1,3-thiazol-4-one
11631681
Crystal-Powder at 20°C

Chemical Identifiers

872573-93-8
351.442
XOLMRFUGOINFDQ-YBEGLDIGSA-N
11631681
C1=CC2=C(C=CC(=C2)C=C3C(=O)N=C(S3)NCC4=CC=CS4)N=C1
C18H13N3OS2
MFCD17392573
cdk1 inhibitor iv, ro-3306, 5-6-quinolinylmethylene-2-2-thienylmethyl amino-4 5h-thiazolone, 5z-5-quinolin-6-ylmethylidene-2-thiophen-2-ylmethyl amino-1,3-thiazol-4-one, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethylamino thiazol-4 5h-one, ro hplc, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethyl amino thiazol-4 5h-one, 2-thiophen-2-ylmethyl-amino-5-1-quinolin-6-yl-meth-z-ylidene-thiazol-4-one, 5z-5-quinolin-6-yl methylidene-2-thiophen-2-yl methyl amino-4,5-dihydro-1,3-thiazol-4-one
(5Z)-5-(quinolin-6-ylmethylidene)-2-(thiophen-2-ylmethylamino)-1,3-thiazol-4-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents