missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RO-3306 95.0+%, TCI America™

$170.20 - $510.63

Chemical Identifiers

CAS 872573-93-8
Molecular Formula C18H13N3OS2
Molecular Weight (g/mol) 351.442
MDL Number MFCD17392573
InChI Key XOLMRFUGOINFDQ-YBEGLDIGSA-N
Synonym cdk1 inhibitor iv, ro-3306, 5-6-quinolinylmethylene-2-2-thienylmethyl amino-4 5h-thiazolone, 5z-5-quinolin-6-ylmethylidene-2-thiophen-2-ylmethyl amino-1,3-thiazol-4-one, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethylamino thiazol-4 5h-one, ro hplc, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethyl amino thiazol-4 5h-one, 2-thiophen-2-ylmethyl-amino-5-1-quinolin-6-yl-meth-z-ylidene-thiazol-4-one, 5z-5-quinolin-6-yl methylidene-2-thiophen-2-yl methyl amino-4,5-dihydro-1,3-thiazol-4-one
PubChem CID 11631681
IUPAC Name (5Z)-5-(quinolin-6-ylmethylidene)-2-(thiophen-2-ylmethylamino)-1,3-thiazol-4-one
SMILES C1=CC2=C(C=CC(=C2)C=C3C(=O)N=C(S3)NCC4=CC=CS4)N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
R020110MG View Documents Tci America
R020110MG
10mg Each for $170.20
 
R020150MG View Documents Tci America
R020150MG
50mg Each for $510.63
 
Specifications

Chemical Identifiers

872573-93-8
351.442
XOLMRFUGOINFDQ-YBEGLDIGSA-N
11631681
C1=CC2=C(C=CC(=C2)C=C3C(=O)N=C(S3)NCC4=CC=CS4)N=C1
C18H13N3OS2
MFCD17392573
cdk1 inhibitor iv, ro-3306, 5-6-quinolinylmethylene-2-2-thienylmethyl amino-4 5h-thiazolone, 5z-5-quinolin-6-ylmethylidene-2-thiophen-2-ylmethyl amino-1,3-thiazol-4-one, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethylamino thiazol-4 5h-one, ro hplc, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethyl amino thiazol-4 5h-one, 2-thiophen-2-ylmethyl-amino-5-1-quinolin-6-yl-meth-z-ylidene-thiazol-4-one, 5z-5-quinolin-6-yl methylidene-2-thiophen-2-yl methyl amino-4,5-dihydro-1,3-thiazol-4-one
(5Z)-5-(quinolin-6-ylmethylidene)-2-(thiophen-2-ylmethylamino)-1,3-thiazol-4-one

Specifications

Undesignated
872573-93-8
MFCD17392573
XOLMRFUGOINFDQ-YBEGLDIGSA-N
(5Z)-5-(quinolin-6-ylmethylidene)-2-(thiophen-2-ylmethylamino)-1,3-thiazol-4-one
11631681
Crystal-Powder at 20°C
RO-3306
10mg
C18H13N3OS2
cdk1 inhibitor iv, ro-3306, 5-6-quinolinylmethylene-2-2-thienylmethyl amino-4 5h-thiazolone, 5z-5-quinolin-6-ylmethylidene-2-thiophen-2-ylmethyl amino-1,3-thiazol-4-one, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethylamino thiazol-4 5h-one, ro hplc, z-5-quinolin-6-ylmethylene-2-thiophen-2-ylmethyl amino thiazol-4 5h-one, 2-thiophen-2-ylmethyl-amino-5-1-quinolin-6-yl-meth-z-ylidene-thiazol-4-one, 5z-5-quinolin-6-yl methylidene-2-thiophen-2-yl methyl amino-4,5-dihydro-1,3-thiazol-4-one
C1=CC2=C(C=CC(=C2)C=C3C(=O)N=C(S3)NCC4=CC=CS4)N=C1
351.442
351.44
≥95.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator