Please login to your online account to display your discounted pricing

Ro 106-9920, Tocris

Inhibitor of NF-κB activation

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material Ro 106-9920
Assay Percent Range >99%
Molecular Formula C10H7N5OS
Formula Weight 245.26
Synonym 6-(Phenylsulfinyl)tetrazolo[1, 5-b]pyridazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

177850

 
r & d systems
177850
50mg Each for $746.48

177810

 
r & d systems
177810
10mg Each for $177.37
Description & Specifications

Specifications

Formula Weight 245.26
Quantity 50mg
Molecular Formula C10H7N5OS
Synonym 6-(Phenylsulfinyl)tetrazolo[1, 5-b]pyridazine
CAS 62645-28-7
Assay Percent Range >99%
Chemical Name or Material Ro 106-9920
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C