Please login to your online account to display your discounted pricing

Ro 10-5824 dihydrochloride, Tocris

Selective D4 receptor partial agonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material Ro 10-5824 dihydrochloride
Assay Percent Range >99%
Molecular Formula C17H20N4·2HCl
Formula Weight 353.29
Synonym 5-[(3, 6-Dihydro-4-phenyl-1(2H)-pyridinyl)methyl]-2-methyl-4-pyrimidinamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

232910

 
r & d systems
232910
10mg Each for $177.37

232950

 
r & d systems
232950
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 353.29
Quantity 10mg
Chemical Name or Material Ro 10-5824 dihydrochloride
Assay Percent Range >99%
Molecular Formula C17H20N4·2HCl
CAS 189744-94-3
Synonym 5-[(3, 6-Dihydro-4-phenyl-1(2H)-pyridinyl)methyl]-2-methyl-4-pyrimidinamine dihydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature