Please login to your online account to display your discounted pricing

Ro 04-6790, Tocris

Potent and selective 5-HT6 antagonist

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Ro 04-6790
Assay Percent Range >98%
Molecular Formula C12H16N6O2S
Formula Weight 308.36
Synonym 4-Amino-N-[2, 6-bis(methylamino)-4-pyrimidinyl]-benzenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

291110

 
r & d systems
RDS291110
10mg Each for $201.36

291150

 
r & d systems
RDS291150
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 308.36
Quantity 10mg
Chemical Name or Material Ro 04-6790
Assay Percent Range >98%
Molecular Formula C12H16N6O2S
CAS 202466-68-0
Synonym 4-Amino-N-[2, 6-bis(methylamino)-4-pyrimidinyl]-benzenesulfonamide
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C