Please login to your online account to display your discounted pricing

Ro 04-5595 hydrochloride, Tocris

Selective NR2B antagonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material Ro 04-5595 hydrochloride
Assay Percent Range >98%
Molecular Formula C19H22ClNO2·HCl
Formula Weight 368.3
Synonym 1-[2-(4-Chlorophenyl)ethyl]-1,2,3,4-tetrahydro-6-methoxy-2-methyl-7-isoquinolinol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

200550

 
r & d systems
RDS200550
50mg Each for $924.23

200510

 
r & d systems
RDS200510
10mg Each for $225.77
Description & Specifications

Specifications

Formula Weight 368.3
Quantity 50mg
Assay Percent Range >98%
CAS 64047-73-0
Chemical Name or Material Ro 04-5595 hydrochloride
Molecular Formula C19H22ClNO2·HCl
Synonym 1-[2-(4-Chlorophenyl)ethyl]-1,2,3,4-tetrahydro-6-methoxy-2-methyl-7-isoquinolinol hydrochloride
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C