Please login to your online account to display your discounted pricing

Ro 01-6128, Tocris™

Positive allosteric modulator of mGlu1

$126.32 - $534.65

Chemical Identifiers

Chemical Name or Material Ro 01-6128
Assay Percent Range >98%
Molecular Formula C17H17NO3
Formula Weight 283.32
Synonym (Diphenylacetyl)-carbamic acid ethy ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

434810

 
r & d systems
RDS434810
10mg Each for $126.32

434850

 
r & d systems
RDS434850
50mg Each for $534.65
Description & Specifications

Specifications

Formula Weight 283.32
Quantity 10mg
CAS 302841-86-7
Molecular Formula C17H17NO3
Recommended Storage Store at +4°C
Chemical Name or Material Ro 01-6128
Assay Percent Range >98%
Synonym (Diphenylacetyl)-carbamic acid ethy ester
Safety and Handling

Store at +4°C