Please login to your online account to display your discounted pricing

Thermo Scientific™ RNase I


Ribonuclease I (RNase I) is an endoribonuclease that hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates.

Manufacturer: thermo scientific™  EN0602

 View more versions of this product

Catalog No. FEREN0602

  • / Each
 

Description & Specifications

Specifications

Source E.coli cells with a cloned rna gene of E.coli
Concentration 10U/µL
Molecular Weight 29.6kDa monomer
Units 5,000U
Inhibitors SDS at 0.1% concentration irreversibly inactivates the enzyme

Ribonuclease I (RNase I), an endoribonuclease, preferentially hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates via nucleoside 2', 3'-cyclic monophosphate intermediates.

The enzyme does not require any metal ions for activity.

Highlights

  • Stable and active under a wide variety of reaction conditions
  • Can be heat inactivated in 30 minutes at 100°C