missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ RNase I

Ribonuclease I (RNase I) is an endoribonuclease that hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates.

Manufacturer:  Thermo Scientific™ EN0601

 View more versions of this product

Catalog No. FEREN0601


 

Specifications

Description

Ribonuclease I (RNase I), an endoribonuclease, preferentially hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates via nucleoside 2', 3'-cyclic monophosphate intermediates.

The enzyme does not require any metal ions for activity.

Highlights

  • Stable and active under a wide variety of reaction conditions
  • Can be heat inactivated in 30 minutes at 100°C
Description & Specifications

Specifications

10U/µL
E.coli cells with a cloned rna gene of E.coli
1,000U
SDS at 0.1% concentration irreversibly inactivates the enzyme
29.6kDa monomer