missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ RNA sample loading buffer, no ethidium bromide

Specifications

Chemical Name or Material RNA sample loading buffer
Name Note no ethidium bromide
MDL Number MFCD00213041
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61937XF View Documents Alfa Aesar™
J61937XF
1mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ61937S0 View Documents Alfa Aesar™
J61937S0
2mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Formulated for electrophoresis of RNA on formaldehyde-agarose gels with or without ethidium bromide.
Specifications

Specifications

RNA sample loading buffer
no ethidium bromide
MFCD00213041