Please login to your online account to display your discounted pricing

RN 1747, Tocris™

Selective TRPV4 agonist

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material RN 1747
Assay Percent Range >98%
Molecular Formula C17H18ClN3O4S
Formula Weight 395.87
Synonym 1-(4-Chloro-2-nitrophenyl)sulfonyl-4-benzylpiperazine
View More Specs
Products
Catalog Number Mfr. No. Description Price Quantity    

374510

 
r & d systems
RDS374510
Each for $168.15

374550

 
r & d systems
RDS374550
Each for $697.40
Description & Specifications

Specifications

Formula Weight 395.87
Quantity 10mg
Assay Percent Range >98%
Molecular Formula C17H18ClN3O4S
CAS 1024448-59-6
Synonym 1-(4-Chloro-2-nitrophenyl)sulfonyl-4-benzylpiperazine
Recommended Storage Store at +4°C
Chemical Name or Material RN 1747
Safety and Handling

Store at +4°C