Please login to your online account to display your discounted pricing

RN 1734, Tocris™

Selective TRPV4 antagonist

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material RN 1734
Assay Percent Range >98%
Molecular Formula C14H22CL2N2O2S
Formula Weight 353.31
Synonym 2, 4-Dichloro-N-isopropyl-N-(2-isopropylaminoethyl)benzenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

374610

 
r & d systems
RDS374610
10mg Each for $168.15

374650

 
r & d systems
RDS374650
50mg Each for $697.40
Description & Specifications

Specifications

Formula Weight 353.31
Quantity 10mg
Chemical Name or Material RN 1734
Assay Percent Range >98%
Molecular Formula C14H22CL2N2O2S
CAS 946387-07-1
Synonym 2, 4-Dichloro-N-isopropyl-N-(2-isopropylaminoethyl)benzenesulfonamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C