Please login to your online account to display your discounted pricing

RJR 2429 dihydrochloride, Tocris


Nicotinic receptor agonist

Quantity 10mg

Manufacturer: r & d systems  RDS127110

 View more versions of this product

Catalog No. 127110

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 261.19
Quantity 10mg
Assay Percent Range >98%
CAS 1021418-53-0
Chemical Name or Material RJR 2429 dihydrochloride
Molecular Formula C12H16N2·2HCl
Synonym (±)-2-(3-Pyridinyl)-1-azabicyclo[2.2.2]octane dihydrochloride
Recommended Storage Desiccate at Room Temperature

Specifications

Chemical Name or Material RJR 2429 dihydrochloride
Assay Percent Range >98%
Molecular Formula C12H16N2·2HCl
Formula Weight 261.19
Synonym (±)-2-(3-Pyridinyl)-1-azabicyclo[2.2.2]octane dihydrochloride
Accessories

Desiccate at Room Temperature