Please login to your online account to display your discounted pricing

RJR 2429 dihydrochloride, Tocris

Nicotinic receptor agonist

$199.70 - $834.54

Chemical Identifiers

Chemical Name or Material RJR 2429 dihydrochloride
Assay Percent Range >98%
Molecular Formula C12H16N2·2HCl
Formula Weight 261.19
Synonym (±)-2-(3-Pyridinyl)-1-azabicyclo[2.2.2]octane dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

127150

 
r & d systems
RDS127150
50mg Each for $834.54

127110

 
r & d systems
RDS127110
10mg Each for $199.70
Description & Specifications

Specifications

Formula Weight 261.19
Quantity 50mg
Chemical Name or Material RJR 2429 dihydrochloride
Assay Percent Range >98%
CAS 1021418-53-0
Synonym (±)-2-(3-Pyridinyl)-1-azabicyclo[2.2.2]octane dihydrochloride
Recommended Storage Desiccate at Room Temperature
Molecular Formula C12H16N2·2HCl
Safety and Handling

Desiccate at Room Temperature